N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N-cyclopentylamine
Supplier: Matrix Scientific
053046-500MG
053046-2.5G
101902-072EA
364.62
USD
101902-072
101902-074
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N-cyclopentylamine
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N-cyclopentylamine
Hazard: Irritant
Formula:
C₁₁H₁₂ClF₃N₂ |
MDL Number:
MFCD11120659 CAS Number: 1041581-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...