6-Chloro-N-cyclopentyl-4-pyrimidinamine
Supplier: Matrix Scientific
053045-500MG
053045-2.5G
101902-068EA
364.62
USD
101902-068
101902-070
6-Chloro-N-cyclopentyl-4-pyrimidinamine
6-Chloro-N-cyclopentyl-4-pyrimidinamine
Hazard: Irritant
Formula:
C₉H₁₂ClN₃ |
MDL Number:
MFCD13562541 CAS Number: 1220016-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...