5-Bromo-N-cyclopentyl-4-methyl-2-pyridinamine

Supplier: Matrix Scientific

053044-500MG 053044-2.5G
101902-064EA 364.62 USD
101902-064 101902-066
5-Bromo-N-cyclopentyl-4-methyl-2-pyridinamine
5-Bromo-N-cyclopentyl-4-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂
MDL Number: MFCD13562540
CAS Number: 1220034-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR