4-(Cyclopentylamino)-3-nitrobenzoic acid
Supplier: Matrix Scientific
053043-500MG
053043-2.5G
101902-060EA
364.62
USD
101902-060
101902-062
4-(Cyclopentylamino)-3-nitrobenzoic acid
4-(Cyclopentylamino)-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₄ |
MDL Number:
MFCD09031721 CAS Number: 691363-42-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...