Ethyl-3-amino-4-(cyclopentylamino)benzoate

Supplier: Matrix Scientific

053042-500MG 053042-2.5G
101902-056EA 364.62 USD
101902-056 101902-058
Ethyl-3-amino-4-(cyclopentylamino)benzoate
Ethyl-3-amino-4-(cyclopentylamino)benzoate

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
MDL Number: MFCD13343642
CAS Number: 885312-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR