3-Amino-4-chloro-N-(3-hydroxypropyl)benzenesulfonamide
Supplier: Matrix Scientific
053041-500MG
053041-2.5G
101902-052EA
364.62
USD
101902-052
101902-054
3-Amino-4-chloro-N-(3-hydroxypropyl)benzenesulfonamide
3-Amino-4-chloro-N-(3-hydroxypropyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₉H₁₃ClN₂O₃S MW: 264.73 g/mol |
MDL Number:
MFCD11642902 CAS Number: 1097827-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...