N-(3-Hydroxypropyl)-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053038-500MG 053038-2.5G
101902-040EA 364.62 USD
101902-040 101902-042
N-(3-Hydroxypropyl)-3-piperidinecarboxamide hydrochloride
N-(3-Hydroxypropyl)-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD13562537
CAS Number: 1220035-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR