N-(3-Hydroxypropyl)-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053037-500MG
053037-2.5G
101902-036EA
364.62
USD
101902-036
101902-038
N-(3-Hydroxypropyl)-2-piperidinecarboxamide hydrochloride
N-(3-Hydroxypropyl)-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O₂ |
MDL Number:
MFCD13562536 CAS Number: 1246172-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...