2-Amino-N-(3-hydroxypropyl)-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

053035-500MG 053035-2.5G
101902-028EA 364.62 USD
101902-028 101902-030
2-Amino-N-(3-hydroxypropyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(3-hydroxypropyl)-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O₂
MW: 258.75 g/mol
MDL Number: MFCD13562534
CAS Number: 1236260-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR