2-Amino-N-(3-hydroxypropyl)-2-methylpropanamide hydrochloride

Supplier: Matrix Scientific

053034-500MG 053034-2.5G
101902-024EA 364.62 USD
101902-024 101902-026
2-Amino-N-(3-hydroxypropyl)-2-methylpropanamide hydrochloride
2-Amino-N-(3-hydroxypropyl)-2-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂
MDL Number: MFCD13562533
CAS Number: 1219957-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR