N-(3-Hydroxypropyl)-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

053031-500MG 053031-2.5G
101902-012EA 364.62 USD
101902-012 101902-014
N-(3-Hydroxypropyl)-2-(methylamino)acetamide hydrochloride
N-(3-Hydroxypropyl)-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂
MDL Number: MFCD13562530
CAS Number: 1220019-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR