2-Amino-N-(3-hydroxypropyl)propanamide hydrochloride
Supplier: Matrix Scientific
053030-500MG
053030-2.5G
101902-008EA
364.62
USD
101902-008
101902-010
2-Amino-N-(3-hydroxypropyl)propanamide hydrochloride
2-Amino-N-(3-hydroxypropyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₆H₁₅ClN₂O₂ |
MDL Number:
MFCD13562529 CAS Number: 1236254-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...