2-Amino-N-(3-hydroxypropyl)acetamide hydrochloride

Supplier: Matrix Scientific

053029-500MG 053029-2.5G
101902-004EA 166.15 USD
101902-004 101902-006
2-Amino-N-(3-hydroxypropyl)acetamide hydrochloride
2-Amino-N-(3-hydroxypropyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O₂
MDL Number: MFCD13562528
CAS Number: 1220034-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR