3-[(6-Chloro-2-pyridinyl)amino]-1-propanol

Supplier: Matrix Scientific

053028-500MG 053028-2.5G
101902-000EA 364.62 USD
101902-000 101902-002
3-[(6-Chloro-2-pyridinyl)amino]-1-propanol
3-[(6-Chloro-2-pyridinyl)amino]-1-propanol

Hazard: Irritant

Formula: C₈H₁₁ClN₂O
MDL Number: MFCD13562527
CAS Number: 29449-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR