3-[(5-Bromo-3-methyl-2-pyridinyl)amino]-1-propanol
Supplier: Matrix Scientific
053026-500MG
053026-2.5G
101901-992EA
364.62
USD
101901-992
101901-994
3-[(5-Bromo-3-methyl-2-pyridinyl)amino]-1-propanol
3-[(5-Bromo-3-methyl-2-pyridinyl)amino]-1-propanol
Hazard: Irritant
Formula:
C₉H₁₃BrN₂O |
MDL Number:
MFCD13562526 CAS Number: 1220034-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...