2-Chloro-N-(3-hydroxypropyl)nicotinamide
Supplier: Matrix Scientific
053025-500MG
053025-2.5G
101901-988EA
364.62
USD
101901-988
101901-990
2-Chloro-N-(3-hydroxypropyl)nicotinamide
2-Chloro-N-(3-hydroxypropyl)nicotinamide
Hazard: Irritant
Formula:
C₉H₁₁ClN₂O₂ |
MDL Number:
MFCD12776304 CAS Number: 1183452-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...