3-[(3-Amino-2-pyridinyl)amino]-1-propanol

Supplier: Matrix Scientific

053024-500MG 053024-2.5G
101901-984EA 364.62 USD
101901-984 101901-986
3-[(3-Amino-2-pyridinyl)amino]-1-propanol
3-[(3-Amino-2-pyridinyl)amino]-1-propanol

Hazard: Irritant

Formula: C₈H₁₃N₃O
MDL Number: MFCD11101083
CAS Number: 1082123-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR