2-[(3-Hydroxypropyl)amino]isonicotinic acid
Supplier: Matrix Scientific
053022-500MG
053022-2.5G
101901-976EA
364.62
USD
101901-976
101901-978
2-[(3-Hydroxypropyl)amino]isonicotinic acid
2-[(3-Hydroxypropyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₉H₁₂N₂O₃ |
MDL Number:
MFCD13562524 CAS Number: 1220019-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...