3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-1-propanol

Supplier: Matrix Scientific

053020-500MG 053020-2.5G
101901-968EA 364.62 USD
101901-968 101901-970
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-1-propanol
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-1-propanol

Hazard: Irritant

Formula: C₉H₁₀ClF₃N₂O
MDL Number: MFCD11123141
CAS Number: 1040006-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR