3-[(6-Chloro-4-pyrimidinyl)amino]-1-propanol

Supplier: Matrix Scientific

053019-500MG 053019-2.5G
101901-964EA 364.62 USD
101901-964 101901-966
3-[(6-Chloro-4-pyrimidinyl)amino]-1-propanol
3-[(6-Chloro-4-pyrimidinyl)amino]-1-propanol

Hazard: Irritant

Formula: C₇H₁₀ClN₃O
MDL Number: MFCD13562522
CAS Number: 55662-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR