4-[(3-Hydroxypropyl)amino]-3-nitrobenzoic acid

Supplier: Matrix Scientific

053017-500MG 053017-2.5G
101901-956EA 364.62 USD
101901-956 101901-958
4-[(3-Hydroxypropyl)amino]-3-nitrobenzoic acid
4-[(3-Hydroxypropyl)amino]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₅
MDL Number: MFCD00228419
CAS Number: 306280-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR