Ethyl-3-amino-4-[(3-hydroxypropyl)amino]benzoate
Supplier: Matrix Scientific
053016-500MG
053016-2.5G
101901-952EA
384.62
USD
101901-952
101901-954
Ethyl-3-amino-4-[(3-hydroxypropyl)amino]benzoate
Ethyl-3-amino-4-[(3-hydroxypropyl)amino]benzoate
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O₃ |
MDL Number:
MFCD13562520 CAS Number: 1220016-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...