3-Amino-4-chloro-N-(2-methoxyethyl)benzenesulfonamide
Supplier: Matrix Scientific
053015-500MG
053015-2.5G
101901-948EA
364.62
USD
101901-948
101901-950
3-Amino-4-chloro-N-(2-methoxyethyl)benzenesulfonamide
3-Amino-4-chloro-N-(2-methoxyethyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₉H₁₃ClN₂O₃S MW: 264.73 g/mol |
MDL Number:
MFCD09900806 CAS Number: 1017458-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...