N-(2-Methoxyethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Supplier: Matrix Scientific

053014-500MG 053014-2.5G
101901-944EA 364.62 USD
101901-944 101901-946
N-(2-Methoxyethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-(2-Methoxyethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₇ClN₄O₂
MW: 260.73 g/mol
MDL Number: MFCD13562519
CAS Number: 1220016-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR