N-(2-Methoxyethyl)-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053013-500MG
053013-2.5G
101901-940EA
364.62
USD
101901-940
101901-942
N-(2-Methoxyethyl)-4-piperidinecarboxamide hydrochloride
N-(2-Methoxyethyl)-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O₂ |
MDL Number:
MFCD13562518 CAS Number: 1220037-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...