N-(2-Methoxyethyl)-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053012-500MG
053012-2.5G
101901-936EA
364.62
USD
101901-936
101901-938
N-(2-Methoxyethyl)-3-piperidinecarboxamide hydrochloride
N-(2-Methoxyethyl)-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O₂ |
MDL Number:
MFCD13562517 CAS Number: 1220019-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...