N-(2-Methoxyethyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053011-500MG 053011-2.5G
101901-932EA 364.62 USD
101901-932 101901-934
N-(2-Methoxyethyl)-2-piperidinecarboxamide hydrochloride
N-(2-Methoxyethyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD13562516
CAS Number: 1236262-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR