2-Amino-N-(2-methoxyethyl)-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

053009-500MG 053009-2.5G
101901-924EA 364.62 USD
101901-924 101901-926
2-Amino-N-(2-methoxyethyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(2-methoxyethyl)-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O₂
MDL Number: MFCD13562514
CAS Number: 1236255-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR