2-Amino-N-(2-methoxyethyl)-2-methylpropanamide hydrochloride

Supplier: Matrix Scientific

053008-500MG 053008-2.5G
101901-920EA 364.62 USD
101901-920 101901-922
2-Amino-N-(2-methoxyethyl)-2-methylpropanamide hydrochloride
2-Amino-N-(2-methoxyethyl)-2-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂
MDL Number: MFCD13562513
CAS Number: 1220028-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR