2-Amino-N-(2-methoxyethyl)-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

053007-500MG 053007-2.5G
101901-916EA 364.62 USD
101901-916 101901-918
2-Amino-N-(2-methoxyethyl)-3-methylbutanamide hydrochloride
2-Amino-N-(2-methoxyethyl)-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₉ClN₂O₂
MDL Number: MFCD13562512
CAS Number: 1236259-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR