3-Amino-N-(2-methoxyethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053006-500MG
053006-2.5G
101901-912EA
364.62
USD
101901-912
101901-914
3-Amino-N-(2-methoxyethyl)propanamide hydrochloride
3-Amino-N-(2-methoxyethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₆H₁₅ClN₂O₂ |
MDL Number:
MFCD13562511 CAS Number: 1220019-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...