N-(2-Methoxyethyl)-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

053005-500MG 053005-2.5G
101901-908EA 364.62 USD
101901-908 101901-910
N-(2-Methoxyethyl)-2-(methylamino)acetamide hydrochloride
N-(2-Methoxyethyl)-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂
MDL Number: MFCD08060120
CAS Number: 1220016-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR