2-Amino-N-(2-methoxyethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053004-500MG
053004-2.5G
101901-904EA
364.62
USD
101901-904
101901-906
2-Amino-N-(2-methoxyethyl)propanamide hydrochloride
2-Amino-N-(2-methoxyethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₆H₁₅ClN₂O₂ |
MDL Number:
MFCD13562510 CAS Number: 1236260-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...