2-Amino-N-(2-methoxyethyl)acetamide hydrochloride

Supplier: Matrix Scientific

053003-500MG 053003-2.5G
101901-900EA 166.15 USD
101901-900 101901-902
2-Amino-N-(2-methoxyethyl)acetamide hydrochloride
2-Amino-N-(2-methoxyethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O₂
MDL Number: MFCD08060118
CAS Number: 1220037-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR