6-Chloro-N-(2-methoxyethyl)-2-pyridinamine

Supplier: Matrix Scientific

053002-500MG 053002-2.5G
101901-896EA 364.62 USD
101901-896 101901-898
6-Chloro-N-(2-methoxyethyl)-2-pyridinamine
6-Chloro-N-(2-methoxyethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₈H₁₁ClN₂O
MW: 186.64 g/mol
MDL Number: MFCD13562509
CAS Number: 1220019-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR