N-(5-Bromo-2-pyridinyl)-N-(2-methoxyethyl)amine

Supplier: Matrix Scientific

053001-500MG 053001-2.5G
101901-892EA 364.62 USD
101901-892 101901-894
N-(5-Bromo-2-pyridinyl)-N-(2-methoxyethyl)amine
N-(5-Bromo-2-pyridinyl)-N-(2-methoxyethyl)amine

Hazard: Irritant

Formula: C₈H₁₁BrN₂O
MDL Number: MFCD11123673
CAS Number: 1005010-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR