N-(5-Bromo-3-methyl-2-pyridinyl)-N-(2-methoxyethyl)amine

Supplier: Matrix Scientific

053000-500MG 053000-2.5G
101901-888EA 364.62 USD
101901-888 101901-890
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(2-methoxyethyl)amine
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(2-methoxyethyl)amine

Hazard: Irritant

Formula: C₉H₁₃BrN₂O
MDL Number: MFCD13562508
CAS Number: 1220028-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR