2-Chloro-N-(2-methoxyethyl)nicotinamide
Supplier: Matrix Scientific
052999-500MG
052999-2.5G
101901-884EA
364.62
USD
101901-884
101901-886
2-Chloro-N-(2-methoxyethyl)nicotinamide
2-Chloro-N-(2-methoxyethyl)nicotinamide
Hazard: Irritant
Formula:
C₉H₁₁ClN₂O₂ |
MDL Number:
MFCD03398036 CAS Number: 547706-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...