N2-(2-Methoxyethyl)-2,3-pyridinediamine
Supplier: Matrix Scientific
052998-500MG
052998-2.5G
101901-880EA
384.62
USD
101901-880
101901-882
N2-(2-Methoxyethyl)-2,3-pyridinediamine
N2-(2-Methoxyethyl)-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₈H₁₃N₃O |
MDL Number:
MFCD10693580 CAS Number: 868628-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...