2-[(2-Methoxyethyl)amino]nicotinic acid
Supplier: Matrix Scientific
052997-500MG
052997-2.5G
101901-876EA
364.62
USD
101901-876
101901-878
2-[(2-Methoxyethyl)amino]nicotinic acid
2-[(2-Methoxyethyl)amino]nicotinic acid
Hazard: Irritant
Formula:
C₉H₁₂N₂O₃ |
MDL Number:
MFCD03114315 CAS Number: 460363-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...