6-Chloro-N-(2-methoxyethyl)-2-pyrazinamine

Supplier: Matrix Scientific

052995-500MG 052995-2.5G
101901-868EA 364.62 USD
101901-868 101901-870
6-Chloro-N-(2-methoxyethyl)-2-pyrazinamine
6-Chloro-N-(2-methoxyethyl)-2-pyrazinamine

Hazard: Irritant

Formula: C₇H₁₀ClN₃O
MDL Number: MFCD13562507
CAS Number: 1220016-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR