6-Chloro-N-(2-methoxyethyl)-4-pyrimidinamine
Supplier: Matrix Scientific
052993-500MG
052993-2.5G
101901-860EA
166.15
USD
101901-860
101901-862
6-Chloro-N-(2-methoxyethyl)-4-pyrimidinamine
6-Chloro-N-(2-methoxyethyl)-4-pyrimidinamine
Hazard: Irritant
Formula:
C₇H₁₀ClN₃O MW: 187.63 g/mol |
MDL Number:
MFCD13562506 CAS Number: 503610-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...