N-(5-Bromo-4-methyl-2-pyridinyl)-N-(2-methoxyethyl)amine

Supplier: Matrix Scientific

052992-500MG 052992-2.5G
101901-856EA 384.62 USD
101901-856 101901-858
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(2-methoxyethyl)amine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(2-methoxyethyl)amine

Hazard: Irritant

Formula: C₉H₁₃BrN₂O
MDL Number: MFCD13562505
CAS Number: 1220028-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR