Ethyl-3-amino-4-[(2-methoxyethyl)amino]benzoate

Supplier: Matrix Scientific

052990-500MG 052990-2.5G
101901-848EA 364.62 USD
101901-848 101901-850
Ethyl-3-amino-4-[(2-methoxyethyl)amino]benzoate
Ethyl-3-amino-4-[(2-methoxyethyl)amino]benzoate

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₃
MDL Number: MFCD13343621
CAS Number: 1219957-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR