3-Amino-4-chloro-N-(3-hydroxybutyl)benzenesulfonamide

Supplier: Matrix Scientific

052989-500MG 052989-2.5G
101901-844EA 364.62 USD
101901-844 101901-846
3-Amino-4-chloro-N-(3-hydroxybutyl)benzenesulfonamide
3-Amino-4-chloro-N-(3-hydroxybutyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O₃S
MW: 278.76 g/mol
MDL Number: MFCD13562504
CAS Number: 1219982-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR