N-(3-Hydroxybutyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052987-500MG 052987-2.5G
101901-836EA 364.62 USD
101901-836 101901-838
N-(3-Hydroxybutyl)-4-piperidinecarboxamide hydrochloride
N-(3-Hydroxybutyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD13562502
CAS Number: 1220019-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR