N-(3-Hydroxybutyl)-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052986-500MG 052986-2.5G
101901-832EA 364.62 USD
101901-832 101901-834
N-(3-Hydroxybutyl)-3-piperidinecarboxamide hydrochloride
N-(3-Hydroxybutyl)-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD13562501
CAS Number: 1220037-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR