N-(3-Hydroxybutyl)-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052984-500MG 052984-2.5G
101901-824EA 364.62 USD
101901-824 101901-826
N-(3-Hydroxybutyl)-2-pyrrolidinecarboxamide hydrochloride
N-(3-Hydroxybutyl)-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD13562499
CAS Number: 1236254-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR