2-Amino-N-(3-hydroxybutyl)-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052983-500MG 052983-2.5G
101901-820EA 364.62 USD
101901-820 101901-822
2-Amino-N-(3-hydroxybutyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(3-hydroxybutyl)-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁ClN₂O₂
MDL Number: MFCD13562498
CAS Number: 1236256-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR