2-Amino-N-(3-hydroxybutyl)-2-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052982-500MG 052982-2.5G
101901-816EA 364.62 USD
101901-816 101901-818
2-Amino-N-(3-hydroxybutyl)-2-methylpropanamide hydrochloride
2-Amino-N-(3-hydroxybutyl)-2-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₉ClN₂O₂
MW: 210.71 g/mol
MDL Number: MFCD13562497
CAS Number: 1220019-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR